2,2,6-Trimethyl-6-vinyltetrahydropyran-3-ol (mixture of isomers) 95%

Lieferant: Apollo Scientific

OR938663-5G
APOSOR938663-5GEA 68.3 EUR
APOSOR938663-5G
2,2,6-Trimethyl-6-vinyltetrahydropyran-3-ol (mixture of isomers) 95%
2,2,6-Trimethyl-6-vinyltetrahydropyran-3-ol (mixture of isomers)
Formel: C₁₀H₁₈O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06200770
CAS-Nummer: 14049-11-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR