1-Butyl-4-methylpyridiniumchlorid 98%

Lieferant: Apollo Scientific

Synonyme: 1-Butyl-4-methylpyridinium Chloride

OR938667-25G
APOSOR938667-25GEA 95.7 EUR
APOSOR938667-25G
1-Butyl-4-methylpyridiniumchlorid 98%
1-Butyl-4-methylpyridinium chloride
Formel: C₁₀H₁₆ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD03095439
CAS-Nummer: 112400-86-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR