5-Phenyl-1,2,4-oxadiazole-3-carboxylic acid 95%

Lieferant: Apollo Scientific

OR938669-1G
APOSOR938669-1GEA 828 EUR
APOSOR938669-1G
5-Phenyl-1,2,4-oxadiazole-3-carboxylic acid 95%
5-Phenyl-1,2,4-oxadiazole-3-carboxylic acid
Formel: C₉H₆N₂O₃
Molecular Weight: 190,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00974531
CAS-Nummer: 37937-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR