Sodium methanethiosulphonate 95%

Lieferant: Apollo Scientific

OR938724-1G OR938724-5G
APOSOR938724-1GEA 0 EUR
APOSOR938724-1G APOSOR938724-5G
Sodium methanethiosulphonate 95%
Sodium methanethiosulphonate
Formel: CH₃NaO₂S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03425630
CAS-Nummer: 1950-85-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR