4,6-Dimethylnicotinonitrile 95%

Lieferant: Apollo Scientific

OR938736-5G
APOSOR938736-5GEA 149 EUR
APOSOR938736-5G
4,6-Dimethylnicotinonitrile 95%
4,6-Dimethylnicotinonitrile
Formel: C₈H₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00234419
CAS-Nummer: 6623-21-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR