(1R,2S)-Cyclobutane-1,2-dicarboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: cis-Cyclobutane-1,2-dicarboxylic acid

OR938740-1G
APOSOR938740-1GEA 89.9 EUR
APOSOR938740-1G
(1R,2S)-Cyclobutane-1,2-dicarboxylic acid 95%
(1R,2S)-Cyclobutane-1,2-dicarboxylic acid
Formel: C₆H₈O₄
Molecular Weight: 144.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00019263
CAS-Nummer: 1461-94-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR