Methylboronic acid mida ester 95%

Lieferant: Apollo Scientific

Synonyme: 4,8-Dimethyldihydro-4l4,8l4-[1,3,2]oxazaborolo[2,3-b][1,3,2]oxazaborole-2,6(3H,5H)-dione

OR938746-1G
APOSOR938746-1GEA 50.9 EUR
APOSOR938746-1G
Methylboronic acid mida ester 95%
Methylboronic acid mida ester
Formel: C₆H₁₀BNO₄
Molecular Weight: 170.959 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11215248
CAS-Nummer: 1104637-40-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR