1H-Pyrrolo[3,2-c]pyridin-4-amine 97%

Lieferant: Apollo Scientific
Achtung

OR938784-250MG
APOSOR938784-250MGEA 120 EUR
APOSOR938784-250MG
1H-Pyrrolo[3,2-c]pyridin-4-amine 97%
1H-Pyrrolo[3,2-c]pyridin-4-amine
Formel: C₇H₇N₃
Molecular Weight: 133,151 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08690132
CAS-Nummer: 60290-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR