1-Ethyl-1,4-diazepane 95%

Lieferant: Apollo Scientific

OR938795-1G
APOSOR938795-1GEA 0 EUR
APOSOR938795-1G
1-Ethyl-1,4-diazepane 95%
1-Ethyl-1,4-diazepane
Formel: C₇H₁₆N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06808650
CAS-Nummer: 3619-73-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR