2-Methyl-5,7-dihydrothieno[3,4-d]pyrimidine 95%

Lieferant: Apollo Scientific

OR938848-1G
APOSOR938848-1GEA 52.2 EUR
APOSOR938848-1G
2-Methyl-5,7-dihydrothieno[3,4-d]pyrimidine 95%
2-Methyl-5,7-dihydrothieno[3,4-d]pyrimidine
Formel: C₇H₈N₂S
Molecular Weight: 152.217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00038619
CAS-Nummer: 36267-71-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR