1-(tert-Butyldimethylsilyloxy)-2-propanone 97%

Lieferant: Apollo Scientific

OR938890-25G
APOSOR938890-25GEA 139 EUR
APOSOR938890-25G
1-(tert-Butyldimethylsilyloxy)-2-propanone 97%
1-(tert-Butyldimethylsilyloxy)-2-propanone
Formel: C₉H₂₀O₂Si
Lagertemperatur: Raumtemperatur
MDL: MFCD01321330
CAS-Nummer: 74685-00-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR