N,N-Diethyl-5-borono-2-methoxybenzolsulfonamid 95%

Lieferant: Apollo Scientific

Synonyme: 3-(N,N-Diethylsulphamoyl)-4-methoxybenzeneboronic acid

OR9389-1G
APOSOR9389-1GEA 263 EUR
APOSOR9389-1G
N,N-Diethyl-5-borono-2-methoxybenzolsulfonamid 95%
N,N-Diethyl-5-borono-2-methoxybenzolsulfonamid
Formel: C₁₁H₁₈BNO₅S
Schmelzpunkt: 110…112 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08056367
CAS-Nummer: 871333-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR