1-(2-Bromethyl)pyrrol 97%

Lieferant: Apollo Scientific

OR938914-1G
APOSOR938914-1GEA 106 EUR
APOSOR938914-1G
1-(2-Bromethyl)pyrrol 97%
1-(2-Bromoethyl)pyrrole
Formel: C₆H₈BrN
Lagertemperatur: Raumtemperatur
MDL: MFCD00191322
CAS-Nummer: 78358-86-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR