(2,4-Dichloro-3-methylphenyl)amine hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: 2,4-Dichloro-3-methylaniline

OR938922-1G
APOSOR938922-1GEA 82.8 EUR
APOSOR938922-1G
(2,4-Dichloro-3-methylphenyl)amine hydrochloride 95%
(2,4-Dichloro-3-methylphenyl)amine hydrochloride
Formel: C₇H₈Cl₃N
Molecular Weight: 212,51 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13186332
CAS-Nummer: 19853-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR