3-(1H-Pyrrol-1-yl)aniline 95%

Lieferant: Apollo Scientific

OR938968-1G
APOSOR938968-1GEA 87 EUR
APOSOR938968-1G
3-(1H-Pyrrol-1-yl)aniline 95%
3-(1H-Pyrrol-1-yl)aniline
Formel: C₁₀H₁₀N₂
Molecular Weight: 158,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03407309
CAS-Nummer: 89353-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR