Quinoxalin-2-amine 95%
Lieferant: Apollo Scientific
OR938970-1G
OR938970-250MG
APOSOR938970-1GEA
114
EUR
APOSOR938970-1G
APOSOR938970-250MG
Quinoxalin-2-amine 95%
Quinoxalin-2-amine
Formel:
C₈H₇N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00094018 CAS-Nummer: 5424-05-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...