(2,6-Dimethylphenyl)acetonitril 95%

Lieferant: Apollo Scientific

OR938974-1G
APOSOR938974-1GEA 69.6 EUR
APOSOR938974-1G
(2,6-Dimethylphenyl)acetonitril 95%
(2,6-Dimethylphenyl)acetonitril
Formel: C₁₀H₁₁N
Molecular Weight: 145,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00060300
CAS-Nummer: 54708-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR