(1,3,5-Trimethyl-1H-pyrazol-4-yl)methanol 95%
Lieferant: Apollo Scientific
OR938981-1G
APOSOR938981-1GEA
632
EUR
APOSOR938981-1G
(1,3,5-Trimethyl-1H-pyrazol-4-yl)methanol 95%
(1,3,5-Trimethyl-1H-pyrazol-4-yl)methanol
Formel:
C₇H₁₂N₂O Molecular Weight: 140,183 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00810197 CAS-Nummer: 18712-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...