trans-4-Propylcyclohexanol 95%

Lieferant: Apollo Scientific

OR939021-1G OR939021-250MG
APOSOR939021-1GEA 229 EUR
APOSOR939021-1G APOSOR939021-250MG
trans-4-Propylcyclohexanol 95%
trans-4-Propylcyclohexanol
Formel: C₉H₁₈O
Lagertemperatur: Raumtemperatur
MDL: MFCD11114411
CAS-Nummer: 77866-58-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR