Ethyl 2-hydroxypyrimidine-5-carboxylate 97%

Lieferant: Apollo Scientific

OR939066-1G
APOSOR939066-1GEA 62.5 EUR
APOSOR939066-1G
Ethyl 2-hydroxypyrimidine-5-carboxylate 97%
Ethyl 2-hydroxypyrimidine-5-carboxylate
Formel: C₇H₈N₂O₃
Molecular Weight: 168,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09991546
CAS-Nummer: 95928-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR