2-Chloro-1,8-naphthyridine 96%

Lieferant: Apollo Scientific
Achtung

OR939109-250MG
APOSOR939109-250MGEA 102 EUR
APOSOR939109-250MG
2-Chloro-1,8-naphthyridine 96%
2-Chloro-1,8-naphthyridine
Formel: C₈H₅ClN₂
Lagertemperatur: Kühlschrank
MDL: MFCD00234390
CAS-Nummer: 15936-10-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR