(S)-1-(Pyridin-4-yl)ethanol 95%

Lieferant: Apollo Scientific

OR939111-5G
APOSOR939111-5GEA 89.9 EUR
APOSOR939111-5G
(S)-1-(Pyridin-4-yl)ethanol 95%
(S)-1-(Pyridin-4-yl)ethanol
Formel: C₇H₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00077866
CAS-Nummer: 54656-96-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR