Ethenesulphonyl chloride 96%
Lieferant: Apollo Scientific
OR939136-1G
OR939136-250MG
APOSOR939136-1GEA
0
EUR
APOSOR939136-1G
APOSOR939136-250MG
Ethenesulphonyl chloride 96%
Ethenesulphonyl chloride
Formel:
C₂H₃ClO₂S Siedepunkt: 30…31 °C (1 mm Hg) Dichte: 1,393 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD19200056 CAS-Nummer: 6608-47-5 UN: 2920 ADR: 8+3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...