2,6-Dimethyl-4-nitropyridine 95%

Lieferant: Apollo Scientific

OR939179-1G
APOSOR939179-1GEA 68.3 EUR
APOSOR939179-1G
2,6-Dimethyl-4-nitropyridine 95%
2,6-Dimethyl-4-nitropyridine
Formel: C₇H₈N₂O₂
Molecular Weight: 152,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234324
CAS-Nummer: 4913-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR