4-(Dimethylamino)butyronitrile 95%

Lieferant: Apollo Scientific

Synonyme: 4-Dimethylaminobutyronitrile, 4-(Dimethylamino)butanenitrile

OR939183-5G
APOSOR939183-5GEA 119 EUR
APOSOR939183-5G
4-(Dimethylamino)butyronitrile 95%
4-(Dimethylamino)butyronitrile
Formel: C₆H₁₂N₂
Molecular Weight: 112.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00019892
CAS-Nummer: 13989-82-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR