2-Methyl-5-(piperidin-1-ylsulfonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: (2-Methyl-5-(piperidin-1-ylsulphonyl)phenyl)boronic acid

OR9392-1G
APOSOR9392-1GEA 376 EUR
APOSOR9392-1G
2-Methyl-5-(piperidin-1-ylsulfonyl)phenylboronsäure 98%
2-Methyl-5-(piperidin-1-ylsulfonyl)phenylboronsäure
Formel: C₁₂H₁₈BNO₄S
Molecular Weight: 283,1500 g/mol
Schmelzpunkt: 132…136 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08056364
CAS-Nummer: 871333-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR