Ethyl 2-(2-cyanophenyl)acetate 95%

Lieferant: Apollo Scientific

OR939210-1G
APOSOR939210-1GEA 21.9 EUR
APOSOR939210-1G
Ethyl 2-(2-cyanophenyl)acetate 95%
Ethyl 2-(2-cyanophenyl)acetate
Formel: C₁₁H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08437615
CAS-Nummer: 67237-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR