3-Acetylbiphenyl 97%

Lieferant: Apollo Scientific

Synonyme: 1-Biphenyl-3-yl-ethanone, 1-([1,1'-Biphenyl]-3-yl)ethanone

OR939300-1G
APOSOR939300-1GEA 115 EUR
APOSOR939300-1G
3-Acetylbiphenyl 97%
3-Acetylbiphenyl
Formel: C₁₄H₁₂O
Molecular Weight: 196,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00048220
CAS-Nummer: 3112-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR