4-Methyl-piperidine-1-sulphonyl chloride 95%

Lieferant: Apollo Scientific

OR939344-5G
APOSOR939344-5GEA 82.8 EUR
APOSOR939344-5G
4-Methyl-piperidine-1-sulphonyl chloride 95%
4-Methyl-piperidine-1-sulphonyl chloride
Formel: C₆H₁₂ClNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD08445024
CAS-Nummer: 41483-70-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR