Mevalonlactonmethacrylat 95%

Lieferant: Apollo Scientific

Synonyme: Methacrylic Acid 4-Methyl-2-oxotetrahydro-2H-pyran-4-yl Ester, 4-Methyl-2-oxotetrahydro-2H-pyran-4-yl Methacrylate

OR939346-1G
APOSOR939346-1GEA 377 EUR
APOSOR939346-1G
Mevalonlactonmethacrylat 95%
Mevalonlactonmethacrylat
Formel: C₁₀H₁₄O₄
Molecular Weight: 198,22 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 177080-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR