Boc-D-N-Me-Phg-OH 97%
Lieferant: Apollo Scientific
Synonyme:
N-Boc-(R)-a-(methylamino)-benzeneacetic acid
OR939352-1G
APOSOR939352-1GEA
40.6
EUR
APOSOR939352-1G
Boc-D-N-Me-Phg-OH 97%
Boc-D-N-Me-Phg-OH
Formel:
C₁₄H₁₉NO₄ Molecular Weight: 265.3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07366768 CAS-Nummer: 30925-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...