Allylmethylcarbonat 98%

Lieferant: Apollo Scientific

OR939355-100G
APOSOR939355-100GEA 162 EUR
APOSOR939355-100G
Allylmethylcarbonat 98%
Allylmethylcarbonat
Formel: C₅H₈O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00134545
CAS-Nummer: 35466-83-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR