3-(4-Hydroxyphenyl)-1-propanol 95%

Lieferant: Apollo Scientific

OR939367-5G
APOSOR939367-5GEA 172 EUR
APOSOR939367-5G
3-(4-Hydroxyphenyl)-1-propanol 95%
3-(4-Hydroxyphenyl)-1-propanol
Formel: C₉H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00002953
CAS-Nummer: 10210-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR