[(1-Ethylpiperidin-4-yl)methyl]amine 95%
Lieferant: Apollo Scientific
Synonyme:
(1-Ethylpiperidin-4-yl)methanamine
OR939382-1G
APOSOR939382-1GEA
299
EUR
APOSOR939382-1G
[(1-Ethylpiperidin-4-yl)methyl]amine 95%
[(1-Ethylpiperidin-4-yl)methyl]amine
Formel:
C₈H₁₈N₂ Molecular Weight: 142,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05022431 CAS-Nummer: 21168-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...