Ethyl-trans-4-hydroxycyclohexanecarboxylate 98%

Lieferant: Apollo Scientific

Synonyme: trans-4-Hydroxycyclohexanecarboxylic Acid Ethyl Ester, trans-Ethyl 4-hydroxycyclohexanecarboxylate

OR939383-25G
APOSOR939383-25GEA 127 EUR
APOSOR939383-25G
Ethyl-trans-4-hydroxycyclohexanecarboxylate 98%
Ethyl-trans-4-hydroxycyclohexanecarboxylate
Formel: C₉H₁₆O₃
Molecular Weight: 172.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16295187
CAS-Nummer: 3618-04-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR