1-(1-Cyclopenten-1-yl)pyrrolidin 97%

Lieferant: Apollo Scientific
Achtung

OR939391-5G
APOSOR939391-5GEA 98.6 EUR
APOSOR939391-5G
1-(1-Cyclopenten-1-yl)pyrrolidin 97%
1-(1-Cyclopenten-1-yl)pyrrolidin
Formel: C₉H₁₅N
Lagertemperatur: Raumtemperatur
MDL: MFCD00003162
CAS-Nummer: 7148-07-4
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR