Methyl(trimethylsilyl)acetat 95%
Lieferant: Apollo Scientific
Synonyme:
(Trimethylsilyl)essigsäuremethylester
OR939403-25G
OR939403-5G
APOSOR939403-25GEA
237
EUR
APOSOR939403-25G
APOSOR939403-5G
Methyl(trimethylsilyl)acetat 95%
Methyl(trimethylsilyl)acetat
Formel:
C₆H₁₄O₂Si Lagertemperatur: Raumtemperatur |
CAS-Nummer:
2916-76-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...