6-Methyl-2-(methylthio)-1H-pyrimidin-4-one 95%

Lieferant: Apollo Scientific

OR939412-25G
APOSOR939412-25GEA 111 EUR
APOSOR939412-25G
6-Methyl-2-(methylthio)-1H-pyrimidin-4-one 95%
6-Methyl-2-(methylthio)-1H-pyrimidin-4-one
Formel: C₆H₈N₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD01022058
CAS-Nummer: 6328-58-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR