Ethyl 1-methyl-1H-pyrazole-5-carboxylate 95%

Lieferant: Apollo Scientific

OR939418-5G
APOSOR939418-5GEA 63.8 EUR
APOSOR939418-5G
Ethyl 1-methyl-1H-pyrazole-5-carboxylate 95%
Ethyl 1-methyl-1H-pyrazole-5-carboxylate
Formel: C₇H₁₀N₂O₂
Molecular Weight: 154.166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08700760
CAS-Nummer: 197079-26-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR