cis-Bicyclo[3.3.0]octan-3,7-dion 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: cis-Tetrahydropentalene-2,5(1H,3H)-dione

OR939423-25G
APOSOR939423-25GEA 109 EUR
APOSOR939423-25G
cis-Bicyclo[3.3.0]octan-3,7-dion 95%
cis-Bicyclo[3.3.0]octan-3,7-dion
Formel: C₈H₁₀O₂
Molecular Weight: 138.1600 g/mol
Schmelzpunkt: 83…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003771
CAS-Nummer: 51716-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR