3-Methoxypropanoyl chloride 95%

Lieferant: Apollo Scientific

OR939440-1G OR939440-250MG OR939440-500MG
APOSOR939440-1GEA 0 EUR
APOSOR939440-1G APOSOR939440-250MG APOSOR939440-500MG
3-Methoxypropanoyl chloride 95%
3-Methoxypropanoyl chloride
Formel: C₄H₇ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03424697
CAS-Nummer: 4244-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR