6-Methyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine 95%
Lieferant: Apollo Scientific
OR939449-1G
APOSOR939449-1GEA
828
EUR
APOSOR939449-1G
6-Methyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine 95%
6-Methyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine
Formel:
C₈H₁₂N₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00805140 CAS-Nummer: 7496-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...