2,6-Dichloro-4-methoxyaniline 95%

Lieferant: Apollo Scientific

OR939452-1G
APOSOR939452-1GEA 95.7 EUR
APOSOR939452-1G
2,6-Dichloro-4-methoxyaniline 95%
2,6-Dichloro-4-methoxyaniline
Formel: C₇H₇Cl₂NO
Molecular Weight: 192.043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00115065
CAS-Nummer: 6480-66-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR