Chloromethyl-2,2,2-trichloroethyl ether 95%

Lieferant: Apollo Scientific

OR939469-5G
APOSOR939469-5GEA 435 EUR
APOSOR939469-5G
Chloromethyl-2,2,2-trichloroethyl ether 95%
Chloromethyl-2,2,2-trichloroethyl ether
Formel: C₃H₄Cl₄O
Molecular Weight: 197,86 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 69573-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR