N,N-Dimethyl-3-borono-4-methylbenzolsulfonamid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(N,N-Dimethylsulphamoyl)-2-methylbenzeneboronic acid, (5-(N,N-Dimethylsulphamoyl)-2-methylphenyl)boronic acid
OR9395-1G
APOSOR9395-1GEA
361
EUR
APOSOR9395-1G
N,N-Dimethyl-3-borono-4-methylbenzolsulfonamid 98%
N,N-Dimethyl-3-borono-4-methylbenzolsulfonamid
Formel:
C₉H₁₄BNO₄S Molecular Weight: 243,0900 g/mol Schmelzpunkt: 86…92 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08056363 CAS-Nummer: 871332-99-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...