Ethyl pyrrolidine-2-carboxylate 95%

Lieferant: Apollo Scientific

OR939678-1G
APOSOR939678-1GEA 754 EUR
APOSOR939678-1G
Ethyl pyrrolidine-2-carboxylate 95%
Ethyl pyrrolidine-2-carboxylate
Formel: C₇H₁₃NO₂
Molecular Weight: 143,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00126468
CAS-Nummer: 60169-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR