(4-Chloro-3-cyanophenyl)boronic acid 97+%

Lieferant: Apollo Scientific
Achtung

OR9397-1G OR9397-5G OR9397-10G
APOSOR9397-1GEA 21.9 EUR
APOSOR9397-1G APOSOR9397-5G APOSOR9397-10G
(4-Chloro-3-cyanophenyl)boronic acid 97+%
(4-Chloro-3-cyanophenyl)boronic acid
Formel: C₇H₅BClNO₂
Schmelzpunkt: 300…304 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08056360
CAS-Nummer: 871332-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR