1-Methyl-1,2-epoxycyclohexane 95%

Lieferant: Apollo Scientific

Synonyme: 1-Methyl-7-oxabicyclo[4.1.0]heptane, 1,2-Epoxy-1-methylcyclohexane

OR939738-5G
APOSOR939738-5GEA 186 EUR
APOSOR939738-5G
1-Methyl-1,2-epoxycyclohexane 95%
1-Methyl-1,2-epoxycyclohexane
Formel: C₇H₁₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD11846121
CAS-Nummer: 1713-33-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR