exo-2-Bromnorbornan

Lieferant: Apollo Scientific

Synonyme: exo-2-Brombicyclo[2.2.1]heptan

OR939766-10G
APOSOR939766-10GEA 289 EUR
APOSOR939766-10G
exo-2-Bromnorbornan
exo-2-Bromnorbornan
Formel: C₇H₁₁Br
Molecular Weight: 175,07 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2534-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR