2-Amino-5-thiazolcarbonsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Aminothiazole-5-carboxylic acid

OR9398-1G
APOSOR9398-1GEA 21.9 EUR
APOSOR9398-1G
2-Amino-5-thiazolcarbonsäure 95%
2-Amino-5-thiazolecarboxylic acid
Formel: C₄H₄N₂O₂S
Molecular Weight: 144,1500 g/mol
Schmelzpunkt: 182…184 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06203554
CAS-Nummer: 40283-46-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR